This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM134 SIRNA Assays ›
Gene Symbol:
Tmem134
Gene Name:
transmembrane protein 134
Gene Aliases:
2410001H17Rik, AI463452
Chromosome Location:
Chr.19: 4125960 - 4132307 on Build GRCm38
UniGene ID:
Mm.276255
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS288992
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem134
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078649.1 NP_001072117.1 6, 7 576
NM_025889.2 NP_080165.1 5, 6 531
GenBank mRNA AI789565.1 - 4, 5 258
AK002220.1 - 5, 6 525
AK008515.1 - 5, 6 515
AK009263.1 - 4, 5 653
AK009745.1 - 5, 6 507
AK010323.1 - 6, 7 551
AK129014.1 - 6, 7 3337
AK166519.1 - 6, 7 579
BC026765.1 - 6, 7 558 4166287

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001078647, NM_001078648, BQ190138, FQ227694, BI278839, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0