This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EMC3 SIRNA Assays ›
Gene Symbol:
Emc3
Gene Name:
ER membrane protein complex subunit 3
Gene Aliases:
0610039A15Rik, AI225901, AW260416, Pob, Tmem111
Chromosome Location:
Chr.6: 113514887 - 113531638 on Build GRCm38
UniGene ID:
Mm.275989
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS287758
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Emc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175101.3 NP_780310.1 8 899
GenBank mRNA AK020058.1 - 8 870
AK075595.1 - 8 900
AK167819.1 - 8 748
AK195081.1 - 2 168
AK199506.1 - 2 168
BC004641.1 - 8 895 3157147

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001008355, FQ220446, BC085846.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein folding in endoplasmic reticulum
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0