This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DIP2A SIRNA Assays ›
Gene Symbol:
Dip2a
Gene Name:
DIP2 disco-interacting protein 2 homolog A (Drosophila)
Gene Aliases:
4931420H10Rik, AI426328, Dip2, Kiaa0184-hp, mKIAA0184
Chromosome Location:
Chr.10: 76263049 - 76345301 on Build GRCm38
UniGene ID:
Mm.33183
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS287623
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dip2a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081419.2 NP_001074888.2 5 605
GenBank mRNA AB093213.2 - 3 275
AK050040.1 - 4 489
AK052899.1 - 5 547
BC068227.1 - 2 244

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
metabolic process
negative regulation of gene expression
regulation of apoptotic process
molecular_function
catalytic activity
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0