This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POLE3 SIRNA Assays ›
Gene Symbol:
Pole3
Gene Name:
polymerase (DNA directed), epsilon 3 (p17 subunit)
Gene Aliases:
1810034K18Rik, YBL1
Chromosome Location:
Chr.4: 62523798 - 62525014 on Build GRCm38
UniGene ID:
Mm.27444
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS286187
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pole3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021498.2 NP_067473.2 2, 3 177
GenBank mRNA AF230806.1 - 2, 3 177
BC024996.1 - 2, 3 177 5253060
BC034004.1 - 185

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

histone H3 acetylation
DNA binding
DNA-directed DNA polymerase activity
protein binding
transferase activity
nucleotidyltransferase activity
sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0