This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ELP2 SIRNA Assays ›
Gene Symbol:
Elp2
Gene Name:
elongator acetyltransferase complex subunit 2
Gene Aliases:
AU023723, Epl2, StIP1, Statip1
Chromosome Location:
Chr.18: 24603923 - 24638830 on Build GRCm38
UniGene ID:
Mm.25298
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS286153
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Elp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021448.2 NP_067423.2 5, 6 553
GenBank mRNA AF291064.1 - 5, 6 519
AK033246.1 - 5, 6 554
AK034457.1 - 5, 6 552
AK044699.1 - 5, 6 530
AK050960.1 - 5, 6 554
AK147826.1 - 554
AK167853.1 - 5, 6 552
AK168370.1 - 5, 6 554
AK177305.1 - 3, 4 317
BC015284.1 - 5, 6 545 4239056
BC051943.1 - 5, 6 545 5686798

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of JAK-STAT cascade
RNA polymerase II core binding
protein binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0