This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIAS1 SIRNA Assays ›
Gene Symbol:
Pias1
Gene Name:
protein inhibitor of activated STAT 1
Gene Aliases:
2900068C24Rik, Ddxbp1, GBP
Chromosome Location:
Chr.9: 62878366 - 62980879 on Build GRCm38
UniGene ID:
Mm.431253
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS285779
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pias1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019663.3 NP_062637.2 10 1217
GenBank mRNA AF077950.1 - 10 1209
AK013753.1 - 6 551
AK075708.1 - 10 1218
AK153871.1 - 10 1222
BC051417.1 - 10 1214 6515027

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106829, AY644724.

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
JAK-STAT cascade
protein sumoylation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of protein sumoylation
regulation of cell proliferation
negative regulation of apoptotic process
fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of smooth muscle cell differentiation
protein-DNA complex assembly
negative regulation of nucleic acid-templated transcription
DNA binding
transcription corepressor activity
protein binding
protein C-terminus binding
transcription factor binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
SUMO transferase activity
enzyme binding
protein domain specific binding
ubiquitin protein ligase binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
SUMO ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0