This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEP170 SIRNA Assays ›
Gene Symbol:
Cep170
Gene Name:
centrosomal protein 170
Gene Aliases:
4933426L22Rik, A330004A13Rik, AI195353
Chromosome Location:
Chr.1: 176733653 - 176814173 on Build GRCm38
UniGene ID:
Mm.269991
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS285247
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep170
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099637.2 NP_001093107.2 12 1921

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003751234, XM_006249682, XM_006249683, XM_006249681, XM_008769812, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm