This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNX3 SIRNA Assays ›
Gene Symbol:
Snx3
Gene Name:
sorting nexin 3
Gene Aliases:
SDP3
Chromosome Location:
Chr.10: 42502054 - 42535369 on Build GRCm38
UniGene ID:
Mm.277870
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS285110
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snx3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017472.4 NP_059500.2 1 375
GenBank mRNA AF062482.1 - 1 173
AK007798.1 - 1 186
AK013183.1 - 1 400
AK145350.1 - 1 178
AK154728.1 - 1 178
AK163420.1 - 1 733
BC116885.1 - 1 219 40126204
BC116889.1 - 1 219 40126208

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

heme biosynthetic process
transport
endocytosis
response to bacterium
membrane invagination
positive regulation of neuron projection development
protein transport
vesicle organization
protein to membrane docking
regulation of Wnt signaling pathway
regulation of cellular protein metabolic process
regulation of intracellular protein transport
transferrin transport
negative regulation of protein catabolic process
hemoglobin biosynthetic process
negative regulation of viral entry into host cell
negative regulation of phagocytosis
negative regulation of protein transport
intralumenal vesicle formation
negative regulation of early endosome to late endosome transport
protein binding
lipid binding
phosphatidylinositol-5-phosphate binding
protein phosphatase binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0