This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other H2-M10.2 SIRNA Assays ›
Gene Symbol:
H2-M10.2
Gene Name:
histocompatibility 2, M region locus 10.2
Gene Aliases:
4.7H
Chromosome Location:
Chr.17: 36284281 - 36286421 on Build GRCm38
UniGene ID:
Mm.246798
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS283502
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H2-M10.2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177923.1 NP_808591.1 3 536
GenBank mRNA AF539598.1 - 3 536
AY211078.1 - 519
BC137985.1 - 3 605 8861004
BC137987.1 - 3 605 8861006

Back To Top

More Information


Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

antigen processing and presentation of peptide antigen via MHC class I
receptor binding
peptide antigen binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0