siRNA Details
Gene Symbol: Foxo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194060.1 | NP_918949.1 | 2 | 836 | ||
GenBank mRNA | AJ571690.1 | - | 2 | 836 | ||
BC086628.1 | - | 2 | 924 | 6810803 |

Gene Symbol: Foxo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194060.1 | NP_918949.1 | 2 | 836 | ||
GenBank mRNA | AJ571690.1 | - | 2 | 836 | ||
BC086628.1 | - | 2 | 924 | 6810803 |