This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ABI3BP SIRNA Assays ›
Gene Symbol:
Abi3bp
Gene Name:
ABI gene family, member 3 (NESH) binding protein
Gene Aliases:
5033411B22Rik, AI506287, BG172926, D930038M13Rik, TARSH, eratin
Chromosome Location:
Chr.16: 56477801 - 56690135 on Build GRCm38
UniGene ID:
Mm.209236
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS282964
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Abi3bp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014399.2 NP_001014421.2 23 2235
NM_001014422.2 NP_001014422.1 31 2805
NM_001014423.2 NP_001014423.1 35 3045
NM_001014424.2 NP_001014424.1 32 2853
NM_178790.4 NP_848905.2 30 2745
GenBank mRNA AB190339.1 - 30 2691
AB190340.1 - 23 2181
AB190341.1 - 31 2751
AB190342.1 - 35 2991
AB190343.1 - 32 2799
BC026627.1 - 8 754
BC145388.1 - 22 2123 9053946
BC150763.1 - 22 2123 9087674

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
glycosaminoglycan binding
heparin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0