This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBALD2 SIRNA Assays ›
Gene Symbol:
Ubald2
Gene Name:
UBA-like domain containing 2
Gene Aliases:
1110014K08Rik, D030012E24Rik, Fam100b
Chromosome Location:
Chr.11: 116434094 - 116439077 on Build GRCm38
UniGene ID:
Mm.258466
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS282675
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ubald2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176902.3 NP_795876.1 3 578
GenBank mRNA AK050732.1 - 3 460
AK144901.1 - 3 580
BC116983.1 - 3 449 40126302
BC119080.1 - 3 450 8733829
BY734584.1 - 3 580

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0