This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYO9A SIRNA Assays ›
Gene Symbol:
Myo9a
Gene Name:
myosin IXa
Gene Aliases:
4732465J09Rik, C130068I12Rik, C230003M11
Chromosome Location:
Chr.9: 59750856 - 59928866 on Build GRCm38
UniGene ID:
Mm.249545
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS282231
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Myo9a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173018.2 NP_766606.2 44 7792
GenBank mRNA AK028873.1 - 14 1741
AK029836.1 - 13 1605
BC046526.1 - 7 854
BC068298.1 - 17 2173
BC096035.1 - 6 801

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_134335, AJ001713.

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
intracellular signal transduction
positive regulation of GTPase activity
nucleotide binding
motor activity
actin binding
GTPase activator activity
ATP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0