This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACSF2 SIRNA Assays ›
Gene Symbol:
Acsf2
Gene Name:
acyl-CoA synthetase family member 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 94557102 - 94601891 on Build GRCm38
UniGene ID:
Mm.386885
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS281933
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acsf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153807.2 NP_722502.1 2 291
GenBank mRNA AK034209.1 - 2 316
AK153582.1 - 2 323
AK169987.1 - 2 322
BC018371.1 - 2 291 4210220
BC063269.1 - 2 376 4159843

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
biological_process
metabolic process
nucleotide binding
molecular_function
catalytic activity
acyl-CoA ligase activity
succinate-CoA ligase activity
ATP binding
3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity
fatty acid ligase activity
ligase activity
3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity
2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity
benzoyl acetate-CoA ligase activity
2,4-dichlorobenzoate-CoA ligase activity
pivalate-CoA ligase activity
cyclopropanecarboxylate-CoA ligase activity
adipate-CoA ligase activity
citronellyl-CoA ligase activity
mentha-1,3-dione-CoA ligase activity
thiophene-2-carboxylate-CoA ligase activity
2,4,4-trimethylpentanoate-CoA ligase activity
cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
branched-chain acyl-CoA synthetase (ADP-forming) activity
aryl-CoA synthetase (ADP-forming) activity
3-hydroxypropionyl-CoA synthetase activity
perillic acid:CoA ligase (ADP-forming) activity
perillic acid:CoA ligase (AMP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity
pristanate-CoA ligase activity
malonyl-CoA synthetase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0