This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TENM2 SIRNA Assays ›
Gene Symbol:
Tenm2
Gene Name:
teneurin transmembrane protein 2
Gene Aliases:
Odz2, Odz3, Ten-2, Ten-m2
Chromosome Location:
Chr.11: 36006656 - 37236808 on Build GRCm38
UniGene ID:
Mm.439889
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS280409
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tenm2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290702.1 NP_001277631.1 21 4047
NM_011856.4 NP_035986.3 22 4140
GenBank mRNA AB025411.1 - 22 4177
BC141361.1 - 21 4048 9055973

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
axon guidance
calcium-mediated signaling using intracellular calcium source
self proteolysis
receptor binding
protein homodimerization activity
protein heterodimerization activity
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0