This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DOCK4 SIRNA Assays ›
Gene Symbol:
Dock4
Gene Name:
dedicator of cytokinesis 4
Gene Aliases:
5330406C03, 6330411N01Rik, AF263288, C030023J22, mKIAA0716
Chromosome Location:
Chr.12: 40445957 - 40846874 on Build GRCm38
UniGene ID:
Mm.341423
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS280226
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dock4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172803.2 NP_766391.2 31 3601
GenBank mRNA AK030389.1 - 31 3616
AK049953.1 - 1 767
AK122353.1 - 31 3583
BC141138.1 - 31 3318 9055750

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
positive regulation of GTPase activity
cell chemotaxis
guanyl-nucleotide exchange factor activity
GTPase activator activity
SH3 domain binding
PDZ domain binding
receptor tyrosine kinase binding
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0