siRNA Details
| Gene Symbol: Dock4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172803.2 | NP_766391.2 | 26 | 3095 | ||
| GenBank mRNA | AK030389.1 | - | 26 | 3110 | ||
| AK122353.1 | - | 26 | 3077 | |||
| BC141138.1 | - | 26 | 2812 | 9055750 | ||

Dock4| Gene Symbol: Dock4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172803.2 | NP_766391.2 | 26 | 3095 | ||
| GenBank mRNA | AK030389.1 | - | 26 | 3110 | ||
| AK122353.1 | - | 26 | 3077 | |||
| BC141138.1 | - | 26 | 2812 | 9055750 | ||