This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CREBL2 SIRNA Assays ›
Gene Symbol:
Crebl2
Gene Name:
cAMP responsive element binding protein-like 2
Gene Aliases:
AI046348, B230205M03
Chromosome Location:
Chr.6: 134828686 - 134858931 on Build GRCm38
UniGene ID:
Mm.267010
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS279704
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crebl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177687.3 NP_808355.1 3 542
GenBank mRNA AK038845.1 - 1 20
AK045467.1 - 3 543

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of glucose import
positive regulation of lipid biosynthetic process
protein stabilization
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0