This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FRMD5 SIRNA Assays ›
Gene Symbol:
Frmd5
Gene Name:
FERM domain containing 5
Gene Aliases:
1500032A09Rik, 5830442F04, A930004K21Rik, R75430
Chromosome Location:
Chr.2: 121545529 - 121807757 on Build GRCm38
UniGene ID:
Mm.121514
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS279307
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Frmd5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172673.3 NP_766261.2 13 1338
GenBank mRNA AK030883.1 - 13 1094
AK044270.1 - 13 1089
AK146383.1 - 12 1218
BC038623.1 - 13 1075
BC057549.1 - 10 867 6416914
BC062889.1 - 13 1295 5698263

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
cytoskeletal protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0