This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IGFN1 SIRNA Assays ›
Gene Symbol:
Igfn1
Gene Name:
immunoglobulin-like and fibronectin type III domain containing 1
Gene Aliases:
9830123M21Rik
Chromosome Location:
Chr.1: 135953205 - 136040842 on Build GRCm38
UniGene ID:
Mm.119675
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS279081
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Igfn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177642.3 NP_808310.2 24 7797
GenBank mRNA BC107027.1 - 2 121 40050318
BC107028.2 - 2 121 40050319
EU429481.1 - 10 4669
EU429482.1 - 9 1536
EU429483.2 - 19 2950
GU290042.1 - 24 7797

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0