This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM105A SIRNA Assays ›
Gene Symbol:
Fam105a
Gene Name:
family with sequence similarity 105, member A
Gene Aliases:
9830126M18
Chromosome Location:
Chr.15: 27655071 - 27681596 on Build GRCm38
UniGene ID:
Mm.266485
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS278692
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam105a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242423.1 NP_001229352.1 7 924
NM_001242424.1 NP_001229353.1 7 924
NM_198301.2 NP_938043.1 6 774
GenBank mRNA AK036522.1 - 7 899
AK134932.1 - 1 22
AK150760.1 - 7 895
AK152120.1 - 7 895
AK152590.1 - 7 890
AK160030.1 - 7 868
AK172362.1 - 7 878
BC052328.1 - 6 730 6311697

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0