siRNA Details
Gene Symbol: Myo15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087906.2 | - | 2 | 272 | ||
AK173262.1 | - | 22 | 2545 | |||
BB072116.2 | - | 4 | 398 |

Gene Symbol: Myo15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087906.2 | - | 2 | 272 | ||
AK173262.1 | - | 22 | 2545 | |||
BB072116.2 | - | 4 | 398 |