This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EVI2B SIRNA Assays ›
Gene Symbol:
Evi2b Gm21975
Gene Name:
ecotropic viral integration site 2b
predicted gene 21975
Gene Aliases:
Evi-2, Evi2
Evi2a-evi2b
Chromosome Location:
Chr.11: 79513385 - 79523762 on Build GRCm38
Chromosome Location:
Chr.11: 79513385 - 79530609 on Build GRCm38
UniGene ID:
Mm.439665
Mm.439665
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID MSS278171
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Evi2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077496.1 NP_001070964.1 2 1091
GenBank mRNA AK039968.1 - 2 1068
AK170476.1 - 2 1053
AK171686.1 - 2 1008
BC017548.1 - 2 1040 4922351
Gene Symbol: Gm21975
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146023.4 NP_666135.1 4 1526

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0