This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other L3MBTL2 SIRNA Assays ›
Gene Symbol:
L3mbtl2
Gene Name:
l(3)mbt-like 2 (Drosophila)
Gene Aliases:
4732493N06Rik, M4mbt
Chromosome Location:
Chr.15: 81663889 - 81688315 on Build GRCm38
UniGene ID:
Mm.280768
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS277940
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: L3mbtl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289711.1 NP_001276640.1 5 667
NM_001289712.1 NP_001276641.1 5 667
NM_145993.5 NP_666105.2 5 667
GenBank mRNA AK029115.1 - 5 612
AK036510.1 - 5 620
AK132012.1 - 5 620
AK141810.1 - 4 1801
AK145560.1 - 5 609
AK146632.1 - 5 623
AK156236.1 - 5 622
AY237000.1 - 5 600
AY237001.1 - 5 600
AY237002.1 - 5 600
BC023933.1 - 5 608 5325131
BC030864.1 - 5 570 4211489

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
chromatin modification
zinc ion binding
methylated histone binding
histone binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0