This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RELA SIRNA Assays ›
Gene Symbol:
Rela
Gene Name:
v-rel reticuloendotheliosis viral oncogene homolog A (avian)
Gene Aliases:
p65
Chromosome Location:
Chr.19: 5637440 - 5648130 on Build GRCm38
UniGene ID:
Mm.249966
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS276820
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rela
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009045.4 NP_033071.1 7 930
GenBank mRNA AK089489.1 - 7 675
AK144565.1 - 7 675
AK154630.1 - 7 675
AK155321.1 - 7 940
AK155889.1 - 7 672
AK157076.1 - 7 662
AK158072.1 - 7 940
BC003818.1 - 7 689 3489295
BC094053.1 - 7 930 3157927
DQ020177.1 - 5 499
M61909.1 - 7 645

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_199267, BC079457, AY307375.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
liver development
hair follicle development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
defense response
inflammatory response
I-kappaB kinase/NF-kappaB signaling
positive regulation of cell proliferation
response to bacterium
organ morphogenesis
response to organic substance
response to UV-B
positive regulation of Schwann cell differentiation
cytokine-mediated signaling pathway
positive regulation of chondrocyte differentiation
response to muramyl dipeptide
response to cytokine
response to muscle stretch
NIK/NF-kappaB signaling
negative regulation of protein catabolic process
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of interleukin-12 biosynthetic process
innate immune response
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of insulin receptor signaling pathway
regulation of inflammatory response
positive regulation of NF-kappaB transcription factor activity
cellular response to hydrogen peroxide
nucleotide-binding oligomerization domain containing 2 signaling pathway
response to interleukin-1
cellular response to lipopolysaccharide
cellular response to nicotine
cellular response to interleukin-1
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to peptide hormone stimulus
negative regulation of NIK/NF-kappaB signaling
positive regulation of miRNA metabolic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
enzyme binding
protein kinase binding
ubiquitin protein ligase binding
protein complex binding
activating transcription factor binding
phosphate ion binding
identical protein binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
NF-kappaB binding
repressing transcription factor binding
ankyrin repeat binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0