This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PITPNM2 SIRNA Assays ›
Gene Symbol:
Pitpnm2
Gene Name:
phosphatidylinositol transfer protein, membrane-associated 2
Gene Aliases:
NIR3, RDGBA2, Rdgb2, mKIAA1457
Chromosome Location:
Chr.5: 124118689 - 124249760 on Build GRCm38
UniGene ID:
Mm.44261
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS276813
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pitpnm2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289472.1 NP_001276401.1 4, 5 755
NM_011256.3 NP_035386.1 5, 6 946
GenBank mRNA AF058693.1 - 4, 5 614
AK129364.1 - 4, 5 755
AK147486.1 - 4, 5 627
BC070452.1 - 4, 5 755 30621943
BC082303.1 - 5, 6 946

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107139, XM_008769227, BC168908.

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
phospholipid binding
lipid binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0