This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTPRO SIRNA Assays ›
Gene Symbol:
Ptpro
Gene Name:
protein tyrosine phosphatase, receptor type, O
Gene Aliases:
D28, GLEPP1, PTPBK, PTPROt, PTPU2, PTPoc, PTPphi, Ptpn15, R-PTP-O
Chromosome Location:
Chr.6: 137252299 - 137464634 on Build GRCm38
UniGene ID:
Mm.186361
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS276634
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptpro
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164401.1 NP_001157873.1 8 1937
NM_011216.3 NP_035346.3 8 1937
GenBank mRNA AF135166.1 - 8 1902
AF295638.1 - 8 1550
BC052743.1 - 8 1771 6834312

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
monocyte chemotaxis
regulation of glomerular filtration
negative regulation of glomerular filtration
protein dephosphorylation
axon guidance
dephosphorylation
lamellipodium assembly
glomerulus development
peptidyl-tyrosine dephosphorylation
slit diaphragm assembly
glomerular visceral epithelial cell differentiation
negative regulation of canonical Wnt signaling pathway
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
hydrolase activity
phosphatase activity
Wnt-protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0