This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRKAR2A SIRNA Assays ›
Gene Symbol:
Prkar2a
Gene Name:
protein kinase, cAMP dependent regulatory, type II alpha
Gene Aliases:
1110061A24Rik, AI317181, AI836829, RII(alpha)
Chromosome Location:
Chr.9: 108692143 - 108749512 on Build GRCm38
UniGene ID:
Mm.253102
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS276464
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prkar2a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008924.2 NP_032950.1 6 885
GenBank mRNA AF533977.1 - 6 885
AK132728.1 - 6 824
AK132824.1 - 6 816
BC075623.1 - 6 856 6854057
BC080276.1 - 6 879 30354403

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
regulation of protein kinase activity
protein heterooligomerization
negative regulation of cAMP-dependent protein kinase activity
nucleotide binding
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
small molecule binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0