This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDCD4 SIRNA Assays ›
Gene Symbol:
Pdcd4
Gene Name:
programmed cell death 4
Gene Aliases:
D19Ucla1, Ma3, Tis
Chromosome Location:
Chr.19: 53892231 - 53929861 on Build GRCm38
UniGene ID:
Mm.1605
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS276253
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdcd4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168491.1 NP_001161963.1 7 1011
NM_001168492.1 NP_001161964.1 7 1011
NM_011050.4 NP_035180.2 7 1099
GenBank mRNA AK134366.1 - 7 1100
AK172654.1 - 7 967
BC055739.1 - 7 946 6414089
CT010188.1 - 6 809
D50465.1 - 6 879
D86344.1 - 7 895

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
cell aging
positive regulation of smooth muscle cell apoptotic process
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
regulation of protein metabolic process
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header