This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MGMT SIRNA Assays ›
Gene Symbol:
Mgmt
Gene Name:
O-6-methylguanine-DNA methyltransferase
Gene Aliases:
AGT, AI267024, Agat
Chromosome Location:
Chr.7: 136894468 - 137128193 on Build GRCm38
UniGene ID:
Mm.440219
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS275746
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mgmt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008598.2 NP_032624.1 5 562
GenBank mRNA AB092489.1 - 4 473
AB092490.1 - 4 473
AK090389.1 - 552
BC031888.1 - 5 531 5372377
BC096557.1 - 5 565 1366095
M84524.1 - 4 488

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA dealkylation involved in DNA repair
cellular response to DNA damage stimulus
methylation
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA repair
negative regulation of cell death
DNA binding
damaged DNA binding
catalytic activity
methylated-DNA-[protein]-cysteine S-methyltransferase activity
calcium ion binding
methyltransferase activity
transferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0