This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNJ16 SIRNA Assays ›
Gene Symbol:
Kcnj16
Gene Name:
potassium inwardly-rectifying channel, subfamily J, member 16
Gene Aliases:
6430410F18Rik, AI132396, Kir5.1
Chromosome Location:
Chr.11: 110968033 - 111027968 on Build GRCm38
UniGene ID:
Mm.30176
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS275186
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnj16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252207.1 NP_001239136.1 3 558
NM_001252208.1 NP_001239137.1 2 535
NM_001252209.2 NP_001239138.1 2 525
NM_001252210.2 NP_001239139.1 2 506
NM_010604.3 NP_034734.3 2 477
GenBank mRNA AB016197.1 - 1 309
AK032192.1 - 2 491
AK045447.1 - 2 481
AK078905.1 - 2 478
AK128942.1 - 2 477
AK140249.1 - 2 488
AY377989.1 - 1 311
BC033442.1 - 3 533 4223640
BC092296.1 - 1 368 5064724

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
potassium ion import
regulation of ion transmembrane transport
cellular potassium ion transport
inward rectifier potassium channel activity
voltage-gated ion channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0