This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INVS SIRNA Assays ›
Gene Symbol:
Invs
Gene Name:
inversin
Gene Aliases:
Nphp2, inv
Chromosome Location:
Chr.4: 48279760 - 48431953 on Build GRCm38
UniGene ID:
Mm.317706
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS275121
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Invs
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281977.1 NP_001268906.1 12 1801
NM_001281978.1 NP_001268907.1 14 2079
NM_010569.4 NP_034699.3 12 1853
GenBank mRNA AF034860.3 - 12 1795
AJ010902.1 - 12 1794
AK041671.1 - 10 1513
AK158766.1 - 14 2038
BC126929.1 - 12 1690 40081431

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107932.

Gene Ontology Categories:

Function(s) Process(es)

kidney development
multicellular organism development
post-embryonic development
Wnt signaling pathway
pancreas development
animal organ development
embryonic heart tube left/right pattern formation
negative regulation of canonical Wnt signaling pathway
protein binding
calmodulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0