This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ENG SIRNA Assays ›
Gene Symbol:
Eng
Gene Name:
endoglin
Gene Aliases:
AI528660, AI662476, CD105, Endo, S-endoglin
Chromosome Location:
Chr.2: 32646532 - 32682669 on Build GRCm38
UniGene ID:
Mm.225297
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS274150
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eng
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146348.1 NP_001139820.1 7 1316
NM_001146350.1 NP_001139822.1 7 1319
NM_007932.2 NP_031958.2 7 1319
GenBank mRNA AK151302.1 - 7 1289
AY679531.1 - 7 1319
BC029080.1 - 7 1233 4168250
EU330531.1 - 2 167
S69407.1 - 8 1184
X77952.1 - 7 1319

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
angiogenesis
patterning of blood vessels
vasculogenesis
response to hypoxia
positive regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
heart looping
positive regulation of systemic arterial blood pressure
cell migration involved in endocardial cushion formation
regulation of transcription, DNA-templated
protein phosphorylation
cell adhesion
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
heart development
positive regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
cell migration
regulation of transforming growth factor beta receptor signaling pathway
central nervous system vasculogenesis
extracellular matrix disassembly
negative regulation of cell migration
positive regulation of BMP signaling pathway
negative regulation of protein autophosphorylation
positive regulation of collagen biosynthetic process
intracellular signal transduction
wound healing
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
smooth muscle tissue development
artery morphogenesis
venous blood vessel morphogenesis
cell motility
negative regulation of nitric-oxide synthase activity
cell chemotaxis
negative regulation of pathway-restricted SMAD protein phosphorylation
extracellular matrix constituent secretion
detection of hypoxia
transforming growth factor beta-activated receptor activity
transforming growth factor beta receptor, cytoplasmic mediator activity
type II transforming growth factor beta receptor binding
protein binding
galactose binding
glycosaminoglycan binding
type I transforming growth factor beta receptor binding
BMP binding
protein homodimerization activity
transforming growth factor beta binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0