This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ENDOG SIRNA Assays ›
Gene Symbol:
Endog
Gene Name:
endonuclease G
Gene Aliases:
-
Chromosome Location:
Chr.2: 30171401 - 30174069 on Build GRCm38
UniGene ID:
Mm.474473
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS274147
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Endog
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007931.1 NP_031957.1 1 282
GenBank mRNA AB012108.1 - 1 282
AK144459.1 - 1 314
BC030177.1 - 1 291 5029633

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
DNA metabolic process
DNA catabolic process
apoptotic DNA fragmentation
response to tumor necrosis factor
positive regulation of apoptotic process
response to antibiotic
nucleic acid phosphodiester bond hydrolysis
positive regulation of hydrogen peroxide-mediated programmed cell death
positive regulation of apoptotic DNA fragmentation
nucleic acid binding
nuclease activity
endonuclease activity
deoxyribonuclease activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0