This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ELN SIRNA Assays ›
Gene Symbol:
Eln
Gene Name:
elastin
Gene Aliases:
AI385707, AI480567, E030024M20Rik
Chromosome Location:
Chr.5: 134702596 - 134747367 on Build GRCm38
UniGene ID:
Mm.275320
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS274125
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eln
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007925.3 NP_031951.2 24, 25 1634
GenBank mRNA AK041860.1 - 24, 25 1635
AK080277.1 - 24, 25 1663
BC034057.1 - 12, 13 1172
BC051649.1 - 24, 25 1703 5346255
U08210.1 - 24, 25 1621

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal muscle tissue development
extracellular matrix organization
regulation of actin filament polymerization
stress fiber assembly
extracellular matrix structural constituent
protein binding
extracellular matrix binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0