This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ECEL1 SIRNA Assays ›
Gene Symbol:
Ecel1
Gene Name:
endothelin converting enzyme-like 1
Gene Aliases:
DINE, xce
Chromosome Location:
Chr.1: 87142874 - 87157085 on Build GRCm38
UniGene ID:
Mm.140765
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS274058
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ecel1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277925.1 NP_001264854.1 10 1811
NM_021306.3 NP_067281.2 10 1803
GenBank mRNA AB026294.1 - 9 1631
AK039467.1 - 10 1804
AK159365.1 - 10 1813
BC057569.1 - 9 1633 6413731

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006245475, Y16188.

Gene Ontology Categories:

Function(s) Process(es)

respiratory system process
proteolysis
neuropeptide signaling pathway
intracellular signal transduction
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0