This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRYGC SIRNA Assays ›
Gene Symbol:
Crygc
Gene Name:
crystallin, gamma C
Gene Aliases:
Cryg-5
Chromosome Location:
Chr.1: 65071525 - 65073532 on Build GRCm38
UniGene ID:
Mm.491115
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS273847
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crygc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082573.2 NP_001076042.1 3 350
NM_007775.2 NP_031801.1 3 347
GenBank mRNA BC056454.1 - 2 326 6824225
CJ066821.1 - 3 348
EF426301.1 - 3 336
EF426302.1 - 3 339
M64544.1 - 2 299

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

eye development
visual perception
camera-type eye development
structural constituent of eye lens

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0