This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RPS6KA3 SIRNA Assays ›
Gene Symbol:
Rps6ka3
Gene Name:
ribosomal protein S6 kinase polypeptide 3
Gene Aliases:
C130006E23, MAPKAPK-1b, MPK-9, Rsk2, S6K-alpha3, p90RSK3, pp90RSK2
Chromosome Location:
Chr.X: 159255782 - 159368244 on Build GRCm38
UniGene ID:
Mm.328476
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272865
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rps6ka3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148945.2 NP_683747.1 6 801
GenBank mRNA AK079102.1 - 894
AK154778.1 - 6 714
AY083469.1 - 6 463
BC150156.1 - 6 466
BC150478.1 - 6 466

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

toll-like receptor signaling pathway
regulation of translation
protein phosphorylation
cell cycle
phosphorylation
response to lipopolysaccharide
intracellular signal transduction
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription from RNA polymerase II promoter
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ribosomal protein S6 kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0