This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2Q2 SIRNA Assays ›
Gene Symbol:
Ube2q2
Gene Name:
ubiquitin-conjugating enzyme E2Q (putative) 2
Gene Aliases:
3010021M21Rik
Chromosome Location:
Chr.9: 55148459 - 55207529 on Build GRCm38
UniGene ID:
Mm.207894
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272713
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2q2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_180600.3 NP_850931.2 10 1184
GenBank mRNA AK076148.1 - 10 1047
AK083216.1 - 10 1177
AK150776.1 - 10 1009
BC029111.1 - 10 1013 2631258

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
protein K48-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header