This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PFDN4 SIRNA Assays ›
Gene Symbol:
Pfdn4
Gene Name:
prefoldin 4
Gene Aliases:
0610025M10Rik, 1810063B06Rik, C-1, C1, Pfd4
Chromosome Location:
Chr.2: 170496428 - 170519070 on Build GRCm38
UniGene ID:
Mm.28808
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272699
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pfdn4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013369.2 NP_001013387.1 3 396
NM_001110152.2 NP_001103622.1 3 464
NM_001199902.1 NP_001186831.1 3 452
GenBank mRNA AK007940.1 - 3 257
AK136249.1 - 3 465
BC048510.1 - 3 236
BC065407.1 - 3 396
BC147673.1 - 3 297 9056646
BC147674.1 - 3 297 9056647
BU503885.1 - 2 235
CB523981.1 - 1 72

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0