This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SBK1 SIRNA Assays ›
Gene Symbol:
Sbk1
Gene Name:
SH3-binding kinase 1
Gene Aliases:
Sbk
Chromosome Location:
Chr.7: 126272619 - 126294999 on Build GRCm38
UniGene ID:
Mm.29660
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272301
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sbk1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145587.2 NP_663562.1 2 622
GenBank mRNA BC024114.1 - 2 363 5098034
BC025837.1 - 2 459 5135723
BC031759.1 - 2 622 4194152

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0