This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MBTD1 SIRNA Assays ›
Gene Symbol:
Mbtd1
Gene Name:
mbt domain containing 1
Gene Aliases:
AA408199, AI194990, hemp
Chromosome Location:
Chr.11: 93885850 - 93946985 on Build GRCm38
UniGene ID:
Mm.210334
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272237
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mbtd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134012.3 NP_598773.2 13 1681
GenBank mRNA AK028503.1 - 6 623
AK033238.1 - 8 4554
AK161836.1 - 1 64
BC020018.1 - 9 992 2646754
BC062907.1 - 13 1681 6813849
BC064014.1 - 15 1785 6335954

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
embryonic skeletal system development
molecular_function
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0