This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXL14 SIRNA Assays ›
Gene Symbol:
Fbxl14
Gene Name:
F-box and leucine-rich repeat protein 14
Gene Aliases:
AW322056, Fbx14l
Chromosome Location:
Chr.6: 119479668 - 119483886 on Build GRCm38
UniGene ID:
Mm.125477
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272100
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxl14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133940.3 NP_598701.2 1 1269
GenBank mRNA AF467463.1 - 1 461
AK036124.1 - 1 1166
AK084506.1 - 1 1156
AK155154.1 - 1 1152
BC006913.1 - 1 858
BC021329.1 - 1 1162 3710945

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0