This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYNPO SIRNA Assays ›
Gene Symbol:
Synpo
Gene Name:
synaptopodin
Gene Aliases:
9030217H17Rik, 9130229N11, 9330140I15Rik, AW046661
Chromosome Location:
Chr.18: 60593981 - 60660174 on Build GRCm38
UniGene ID:
Mm.252321
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS270579
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Synpo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020250.1 - 3 199

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0