This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNAJC3 SIRNA Assays ›
Gene Symbol:
Dnajc3
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 3
Gene Aliases:
AA408985, AU067833, Dnajc3a, Dnajc3b, Prkri, mp58, p58, p58IPK
Chromosome Location:
Chr.14: 118937932 - 118981702 on Build GRCm38
UniGene ID:
Mm.12616
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS270018
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnajc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008929.3 NP_032955.2 3 369
GenBank mRNA AK146097.1 - 3 333
AK148270.1 - 3 326
BB618718.1 - 3 369
BC013766.1 - 3 314 2646147
U28423.1 - 3 312

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
response to unfolded protein
endoplasmic reticulum unfolded protein response
negative regulation of apoptotic process
proteolysis involved in cellular protein catabolic process
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
protein kinase inhibitor activity
protein binding
chaperone binding
misfolded protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0