This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other METAP2 SIRNA Assays ›
Gene Symbol:
Metap2
Gene Name:
methionine aminopeptidase 2
Gene Aliases:
4930584B20Rik, A930035J23Rik, AI047573, AL024412, AU014659, Amp2, Mnpep, p67, p67eIF2
Chromosome Location:
Chr.10: 93858489 - 93897090 on Build GRCm38
UniGene ID:
Mm.289329
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS249984
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Metap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019648.3 NP_062622.1 6 1224
GenBank mRNA AB003144.1 - 5 535
AF434712.1 - 5 614
AK076804.1 - 5 567
AK090323.1 - 5 555
AK147097.1 - 5 659
AK163318.1 - 5 641
AK165301.1 - 6 1226
AK165845.1 - 5 550
BC002213.1 - 5 636 3490341
BC092052.1 - 5 601 3670685

Back To Top

More Information


Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein processing
peptidyl-methionine modification
N-terminal protein amino acid modification
aminopeptidase activity
protein binding
peptidase activity
metalloexopeptidase activity
hydrolase activity
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0