This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DGCR14 SIRNA Assays ›
Gene Symbol:
Dgcr14
Gene Name:
DiGeorge syndrome critical region gene 14
Gene Aliases:
AI462402, D16H22S1269E, Dgcr1, Dgsi, ES2, Es2el
Chromosome Location:
Chr.16: 17900709 - 17911364 on Build GRCm38
UniGene ID:
Mm.256480
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS248755
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dgcr14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081633.1 NP_001075102.1 6 769
NM_022408.2 NP_071853.2 6 772
GenBank mRNA AF037256.1 - 6 771
AK088520.1 - 6 734
AK142684.1 - 6 737
AK148404.1 - 6 740
BC013711.1 - 6 748 3708646

Back To Top

More Information


Gene Ontology Categories:

Function(s)

mRNA processing
RNA splicing

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0