This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNE2 SIRNA Assays ›
Gene Symbol:
Kcne2
Gene Name:
potassium voltage-gated channel, Isk-related subfamily, gene 2
Gene Aliases:
2200002I16Rik, AW048273, MiRP1
Chromosome Location:
Chr.16: 92292389 - 92298137 on Build GRCm38
UniGene ID:
Mm.156736
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS248368
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcne2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134110.3 NP_598871.1 2 161
GenBank mRNA AK008619.1 - 2 162
BC022699.1 - 2 103 4481325

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
potassium ion import
cellular protein localization
regulation of ion transmembrane transport
cellular response to drug
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
potassium ion export
potassium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
membrane repolarization
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
regulation of potassium ion transmembrane transport
positive regulation of voltage-gated calcium channel activity
positive regulation of proteasomal protein catabolic process
regulation of inward rectifier potassium channel activity
regulation of cyclic nucleotide-gated ion channel activity
regulation of delayed rectifier potassium channel activity
negative regulation of delayed rectifier potassium channel activity
negative regulation of voltage-gated potassium channel activity
inward rectifier potassium channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
protein binding
potassium channel regulator activity
protein homodimerization activity
ion channel binding
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0