This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYT16 SIRNA Assays ›
Gene Symbol:
Syt16
Gene Name:
synaptotagmin XVI
Gene Aliases:
6430703N11, Strep14, Syt14l, syt14r
Chromosome Location:
Chr.12: 73997608 - 74267916 on Build GRCm38
UniGene ID:
Mm.311393
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS248255
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Syt16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172804.2 NP_766392.2 7 1573
GenBank mRNA AB102950.1 - 6 1905
AK032606.1 - 8 2352
AK137129.1 - 7 1581
BC056932.1 - 6 1683 5708126

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

exocytosis
biological_process
synaptic vesicle exocytosis
calcium ion binding
protein binding
phospholipid binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0