This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM150A SIRNA Assays ›
Gene Symbol:
Tmem150a
Gene Name:
transmembrane protein 150A
Gene Aliases:
BC014685, Tmem150
Chromosome Location:
Chr.6: 72355483 - 72359762 on Build GRCm38
UniGene ID:
Mm.491165
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS248158
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem150a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144916.3 NP_659165.1 7 590
GenBank mRNA BC014685.1 - 6 506 4241375
BC028825.1 - 7 592 5097962

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
regulation of autophagy
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0